missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GP2 Protein

A cDNA sequence encoding the GP2 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12031-2ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15919148

  • 131.00€ / 2µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

GP2 Protein
Research Use Only
Recombinant Protein
Baculovirus
MPHLMERMVG SGLLWLALVS CILTQASAVQ RGYGNPIEAS SYGLDLDCGA PGTPEAHVCF DPCQNYTLLD EPFRSTENSA GSQGCDKNMS GWYRFVGEGG VRMSETCVQV HRCQTDAPMW LNGTHPALGD GITNHTACAH WSGNCCFWKT EVLVKACPGG YHVYRLEGTP WCNLRYCTVP RDPSTVEDKC EKACRPEEEC LALNSTWGCF CRQDLNSSDV HSLQPQLDCG PREIKVKVDK CLLGGLGLGE EVIAYLRDPN CSSILQTEER NWVSVTSPVQ ASACRNILER NQTHAIYKNT LSLVNDFIIR DTILNINFQC AYPLDMKVSL QAALQPIVSS LNVSVDGNGE FIVRMALFQD QNYTNPYEGD AVELSVESVL YVGAILEQGD TSRFNLVLRN CYATPTEDKA DLVKYFIIRN SCSNQRDSTI HVEENGQSSE SRFSVQMFMF AGHYDLVFLH CEIHLCDSLN EQCQPSCSRS QVRSEVPAID LARVLDLGPI TRRGAQSPGV MNGTPSTAGF LVAWPMVLLT VLLAWLF
Greater than 95% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GP2 solution is supplied in 16mM HEPES buffer pH 7.5, 80mM NaCl and 20% glycerol.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions