Learn More
enQuireBio™ Recombinant Human GPNMB Protein
A cDNA sequence encoding the GPNMB was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12041-HIS-HEK-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
GPNMB Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
HEK 293 | |
AKRFHDVLGN ERPSAYMREH NQLNGWSSDE NDWNEKLYPV WKRGDMRWKN SWKGGRVQAV LTSDSPALVG SNITFAVNLI FPRCQKEDAN GNIVYEKNCR NEAGLSADPY VYNWTAWSED SDGENGTGQS HHNVFPDGKP FPHHPGWRRW NFIYVFHTLG QYFQKLGRCS VRVSVNTANV TLGPQLMEVT VYRRHGRAYV PIAQVKDVYV VTDQIPVFVT MFQKNDRNSS DETFLKDLPI MFDVLIHDPS HFLNYSTINY KWSFGDNTGL FVSTNHTVNH TYVLNGTFSL NLTVKAAAPG PCPPPPPPPR PSKPTPSLAT TLKSYDSNTP GPAGDNPLEL SRIPDENCQI NRYGHFQATI TIVEGILEVN IIQMTDVLMP VPWPESSLID FVVTCQGSIP TEVCTIISDP TCEITQNTVC SPVDVDEMCL LTVRRTFNGS GTYCVNLTLG DDTSLALTST LISVP KLHHH HHHHHHH |