missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HPGD Protein
A cDNA sequence encoding the HPGD was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12301-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
3248 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MHVNGKVALV TGAAQGIGRA FAEALLLKGA KVALVDWNLE AGVQCKAALD EQFEPQKTLF IQCDVADQQQ LRDTFRKVVD HFGRLDILVN NAGVNNEKNW EKTLQINLVS VISGTYLGLD YMSKQNGGEG GIIINMSSLA GLMPVAQQPV YCASKHGIVG FTRSAALAAN LMNSGVRLNA ICPGFVNTAI LESIEKEENM GQYIEYKDHI KDMIKYYGIL DPPLIANGLI TLIEDDALNG AIMKITTSKG IHFQDYDTTP FQAKTQ | |
Greater than 95% as determined by SDS-PAGE. |
HPGD Protein | |
Research Use Only | |
HPGD | |
Human | |
His | |
The HPGD protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 0.1M NaCl,1mM DTT and 20% glycerol. |