missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSD17B8 Protein
A cDNA sequence encoding the HSD17B8 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12321-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
7923 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MASQLQNRLR SALALVTGAG SGIGRAVSVR LAGEGATVAA CDLDRAAAQE TVRLLGGPGS KEGPPRGNHA AFQADVSEAR AARCLLEQVQ ACFSRPPSVV VSCAGITQDE FLLHMSEDDW DKVIAVNLKG TFLVTQAAAQ ALVSNGCRGS IINISSIVGK VGNVGQTNYA ASKAGVIGLT QTAARELGRH GIRCNSVLPG FIATPMTQKV PQKVVDKITE MIPMGHLGDP EDVADVVAFL ASEDSGYITG TSVEVTGGLF M | |
Greater than 95.0% as determined by SDS-PAGE. |
HSD17B8 Protein | |
Research Use Only | |
HSD17B8 | |
Human | |
His | |
HSD17B8 solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 40% glycerol and 150mM NaCl. |