missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HYKK Protein

A cDNA sequence encoding the HYKK was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12362-2ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15920179

  • 131.00€ / 2µg
Expédition estimée: 28-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

HYKK Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSSGNYQ QSEALSKPTF SEEQASALVE SVFGLKVSKV RPLPSYDDQN FHVYVSKTKD GPTEYVLKIS NTKASKNPDL IEVQNHIIMF LKAAGFPTAS VCHTKGDNTA SLVSVDSGSE IKSYLVRLLT YLPGRPIAEL PVSPQLLYEI GKLAAKLDKT LQRFHHPKLS SLHRENFIWN LKNVPLLEKY LYALGQNRNR EIVEHVIHLF KEEVMTKLSH FRECINHGDL NDHNILIESS KSASGNAEYQ VSGILDFGDM SYGYYVFEVA ITIMYMMIES KSPIQVGGHV LAGFESITPL TAVEKGALFL LVCSRFCQSL VMAAYSCQLY PENKDYLMVT AKTGWKHLQQ MFDMGQKAVE EIWFETAKSY ESGISM
Greater than 90% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HYKK solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4), 40% glycerol, 1mM DTT and 0.1mM PMSF.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions