missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IGFBP 1 Protein

A cDNA sequence encoding the IGFBP 1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10683-5ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15904648

  • 131.00€ / 5µg
Expédition estimée: 04-07-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

IGFBP 1 Protein
Research Use Only
The ED50, as determined by the inhibition of rHuIGF-I-induced proliferation of human MCF-7 cells, is less than 4microg/ml.
Human
Untagged
IGFBP-1 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS.
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Human
APWQCAPCSA EKLALCPPVS ASCSEVTRSA GCGCCPMCAL PLGAACGVAT ARCARGLSCR ALPGEQQPLH ALTRGQGACV QESDASAPHA AEAGSPESPE STEITEEELL DNFHLMAPSE EDHSILWDAI STYDGSKALH VTNIKKWKEP CRIELYRVVE SLAKAQETSG EEISKFYLPN CNKNGFYHSR QCETSMDGEA GLCWCVYPWN GKRIPGSPEI RGDPNCQIYF NVQN
Greater than 95.0% as determined by SDS-PAGE.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions