Learn More
enQuireBio™ Recombinant Human IL-9 / Interleukin-9 Protein
Recombinant Protein
Marque: enQuireBio™ QP5384-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_000581.1 | |
IL-9 / Interleukin-9 Protein | |
Research Use Only | |
IL9 | |
Recombinant Protein | |
E. coli | |
MQGCPTLAGI LDINFLINKM QEDPASKCHC SANVTSCLCL GIPSDNCTRP CFSERLSQMT NTTMQTRYPL IFSRVKKSVE VLKNNKCPYF SCEQPCNQTT AGNALTFLKS LLEIFQKEKM RGMRGKI | |
Greater than 95% as determined by SDS-PAGE. |
3578 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 2 ng/mL in the MO7e cell proliferation assay. The bioactivity acceptance criteria is set at 5 x 10^5 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |