Learn More
enQuireBio™ Recombinant Human L-lactate dehydrogenase C chain Protein
A DNA sequence encoding the Homo sapiens (Human) L-lactate dehydrogenase C chain, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6291-ba-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_002292.1 | |
L-lactate dehydrogenase C chain Protein | |
20 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3948 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
LDHC | |
Recombinant Protein | |
Baculovirus | |
STVKEQLIEK LIEDDENSQC KITIVGTGAV GMACAISILL KDLADELALV DVALDKLKGE MMDLQHGSLF FSTSKITSGK DYSVSANSRI VIVTAGARQQ EGETRLALVQ RNVAIMKSII PAIVHYSPDC KILVVSNPVD ILTYIVWKIS GLPVTRVIGS GCNLDSARFR YLIGEKLGVH PTSCHGWIIG EHGDSSVPLW SGVNVAGVAL KTLDPKLGTD SDKEHWKNIH KQVIQSAYEI IKLKGYTSWA IGLSVMDLVG SILKNLRRVH PVSTMVKGLY GIKEELFLSI PCVLGRNGVS DVVKINLNSE EEALFKKSAE TLWNIQKDLI F |