Learn More
enQuireBio™ Recombinant Human LAMTOR2 Protein
A cDNA sequence encoding the LAMTOR2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12533-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
LAMTOR2 Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
LAMTOR2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLRPKAL TQVLSQANTG GVQSTLLLNN EGSLLAYSGY GDTDARVTAA IASNIWAAYD RNGNQAFNED NLKFILMDCM EGRVAITRVA NLLLCMYAKE TVGFGMLKAK AQALVQYLEE PLTQVAAS |