Learn More
enQuireBio™ Recombinant Human MRPL48 Protein
A cDNA sequence encoding the MRPL48 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12731-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
MRPL48 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MRPL48 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSGEKPIY SVGGILLSIS RPYKTKPTHG IGKYKHLIKA EEPKKKKGKV EVRAINLGTD YEYGVLNIHL TAYDMTLAES YAQYVHNLCN SLSIKVEESY AMPTKTIEVL QLQDQGSKML LDSVLTTHER VVQISGLSAT FAEIFLEIIQ SSLPEGVRLS VKEHTEEDFK GRFKARPELE ELLAKLK |