missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MVK Protein
A cDNA sequence encoding the MVK was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12760-1mg
Code nomenclature Nacres: NA.28
Informations supplémentaires : Poids : 0.01000kg
Spécification
MVK Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLSEVLL VSAPGKVILH GEHAVVHGKV ALAVSLNLRT FLRLQPHSNG KVDLSLPNIG IKRAWDVARL QSLDTSFLEQ GDVTTPTSEQ VEKLKEVAGL PDDCAVTERL AVLAFLYLYL SICRKQRALP SLDIVVWSEL PPGAGLGSSA AYSVCLAAAL LTVCEEIPNP LKDGDCVNRW TKEDLELINK WAFQGERMIH GNPSGVDNAV STWGGALRYH QGKISSLKRS PALQILLTNT KVPRNTRALV AGVRNRLLKF PEIVAPLLTS IDAISLECER VLGEMGEAPA PEQYLVLEEL IDMNQHHLNA LGVGHASLDQ LCQVTRARGL HSKLTGAGGG GCGITLLKPG LEQPEVEATK QALTSCGFDC LETSIGAPGV SIHSATSLDS RVQQALDGL | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MVK protein (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |