missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDRG3 Protein
A cDNA sequence encoding the NDRG3 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12810-1mg
Informations supplémentaires : Poids : 0.01000kg
Specifications
NDRG3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDELQDV QLTEIKPLLN DKEHDIETTH GVVHVTIRGL PKGNRPVILT YHDIGLNHKS CFNAFFNFED MQEITQHFAV CHVDAPGQQE GAPSFPTGYQ YPTMDELAEM LPPVLTHLSL KSIIGIGVGA GAYILSRFAL NHPELVEGLV LINVDPCAKG WIDWAASKLS GLTTNVVDII LAHHFGQEEL QANLDLIQTY RMHIAQDINQ DNLQLFLNSY NGRRDLEIER PILGQNDNKS KTLKCSTLLV VGDNSPAVEA VVECNSRLNP INTTLLKMAD CGGLPQVVQP GKLTEAFKYF LQGMGYIPSA SMTRLARSRT HSTSSSLGSG ESPFSRSVTS NQSDGTQESC ESPDVLDRHQ TMEVSC | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NDRG3 solution (0.25 mg/1ml) contains Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |