Learn More
enQuireBio™ Recombinant Human NPPA Protein
A cDNA sequence encoding the NPPA was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12873-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
4878 | |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
NPPA | |
Human | |
His | |
NPPA filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 20 mM Tris buffer, 50 mM NaCl and 5% w/v trehalosa, pH 7.5. |
NPPA Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKHHHHHHNP MYNAVSNADL MDFKNLLDHL EEKMPLEDEV VPPQVLSEPN EEAGAALSPL PEVPPWTGEV SPAQRDGGAL GRGPWDSSDR SALLKSKLRA LLTAPR |