Learn More
enQuireBio™ Recombinant Human Nuclear receptor ROR-gamma Protein
A DNA sequence encoding the Homo sapiens (Human) Nuclear receptor ROR-gamma, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6616-ec-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001001523.1 | |
Nuclear receptor ROR-gamma Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6097 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
RORC | |
Recombinant Protein | |
E. coli | |
MDRAPQRQHR ASRELLAAKK THTSQIEVIP CKICGDKSSG IHYGVITCEG CKGFFRRSQR CNAAYSCTRQ QNCPIDRTSR NRCQHCRLQK CLALGMSRDA VKFGRMSKKQ RDSLHAEVQK QLQQRQQQQQ EPVVKTPPAG AQGADTLTYT LGLPDGQLPL GSSPDLPEAS ACPPGLLKAS GSGPSYSNNL AKAGLNGASC HLEYSPERGK AEGRESFYST GSQLTPDRCG LRFEEHRHPG LGELGQGPDS YGSPSFRSTP EAPYASLTEI EHLVQSVCKS YRETCQLRLE DLLRQRSNIF SREEVTGYQR KSMWEMWERC AHHLTEAIQY VVEFAKRLSG FMELCQNDQI VLLKAGAMEV VLVRMCRAYN ADNRTVFFEG KYGGMELFRA LGCSELISSI FDFSHSLSAL HFSEDEIALY TALVLINAHR PGLQEKRKVE QLQYNLELAF HHHLCKTHRQ SILAKLPPKG KLRSLCSQHV ERLQIFQHLH PIVVQAAFPP LYKELFSTET ESPVGLSK |