Learn More
enQuireBio™ Recombinant Human PDGFA Protein
Recombinant Protein
Marque:  enQuireBio™ QP5283-1mg
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15995839
  • 5220.00€ / 1mg
Expédition estimée: 10-05-2024
Spécification
NP_002598.4
PDGFA Protein
Research Use Only
PDGFA
Recombinant Protein
E. coli
MSIEEAVPAV CKTRTVIYEI PRSQVDPTSA NFLIWPPCVE VKRCTGCCNT SSVKCQPSRV HHRSVKVAKV EYVRKKPKLK EVQVRLEEHL ECACATTSLN PDYREEDTGR PRESGKKRKR KRLKPT
Greater than 95% as determined by SDS-PAGE.
5154
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 50 ng/mL in the BALB/c 3T3 cell proliferation assay. The bioactivity acceptance criteria is set at 2.0 x 10^4 units/mg.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA