Learn More
enQuireBio™ Recombinant Human PI3Kb Protein
A cDNA sequence encoding the PI3Kb was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10818-2.5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
PI3Kb Protein | |
2.5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Insect | |
0.9 mg/ml solution in PBS and 2mM MgCl2. |
Greater than 90.0% as determined by SDS Page. | |
Research Use Only | |
The specific activity was found to be 3,000 units/mg (1 unit is defined as 1 picomole phosphate transferred to PIP2 per minute). | |
Human | |
Untagged |