missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PIN4 Protein
A cDNA sequence encoding the PIN4 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10821-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
5303 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 300 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-HCl pH8.0 using chymotrypsin. | |
Human | |
His | |
PIN4 solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 0.1mM PMSF. |
PIN4 Protein | |
Research Use Only | |
PIN4 | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPMAGLLKGL VRQLERFSVQ QQASKMPPKG KSGSGKAGKG GAASGSDSAD KKAQGPKGGG NAVKVRHILC EKHGKIMEAM EKLKSGMRFN EVAAQYSEDK ARQGGDLGWM TRGSMVGPFQ EAAFALPVSG MDKPVFTDPP VKTKFGYHII MVEGRK | |
Greater than 85.0% as determined by SDS-PAGE. |