missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PLA2G1B Protein

A cDNA sequence encoding the PLA2G1B was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13072-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15952419

  • 5655.00€ / 1mg
Expédition estimée: 25-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

PLA2G1B Protein
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASHMAVWQ FRKMIKCVIP GSDPFLEYNN YGCYCGLGGS GTPVDELDKC CQTHDNCYDQ AKKLDSCKFL LDNPYTHTYS YSCSGSAITC SSKNKECEAF ICNCDRNAAI CFSKAPYNKA HKNLDTKKYC QS
Greater than 95% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions