Learn More
enQuireBio™ Recombinant Human Pleiotrophin / PTN / HB-GAM Protein
Recombinant Protein
Marque:  enQuireBio™ QP5365-1mg
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15995989
  • 4210.00€ / 1mg
Expédition estimée: 09-08-2022
Spécification
NP_001308315.1
Pleiotrophin / PTN / HB-GAM Protein
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MGKKEKPEKK VKKSDCGEWQ WSVCVPTSGD CGLGTREGTR TGAECKQTMK TQRCKIPCNW KKQFGAECKY QFQAWGECDL NTALKTRTGS LKRALHNAEC QKTVTISKPC GKLTKPKPQA ESKKKKKEGK KQEKMLD
5764
Greater than 95% as determined by SDS-PAGE.
Research Use Only
PTN
Human
Untagged
10 mM sodium phosphate, pH 7.5