missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Plexin-A1 Protein
A DNA sequence encoding the Homo sapiens (Human) Plexin-A1, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP8791-ye-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_115618.3 | |
Plexin-A1 Protein | |
Research Use Only | |
PLXNA1 | |
Recombinant Protein | |
Yeast | |
LNAGSDVAVS VGGRPCSFSW RNSREIRCLT PPGQSPGSAP IIININRAQL TNPEVKYNYT EDPTILRIDP EWSINSGGTL LTVTGTNLAT VREPRIRAKY GGIERENGCL VYNDTTMVCR APSVANPVRS PPELGERPDE LGFVMDNVRS LLVLNSTSFL YYPDPVL | |
Greater than 90% as determined by SDS-PAGE. |
5361 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |