missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Potassium-transporting ATPase subunit beta Protein
A DNA sequence encoding the Homo sapiens (Human) Potassium-transporting ATPase subunit beta, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5692-ec-200ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_000696.1 | |
Potassium-transporting ATPase subunit beta Protein | |
Research Use Only | |
ATP4B | |
Recombinant Protein | |
E. coli | |
CLYVLMQTVD PYTPDYQDQL RSPGVTLRPD VYGEKGLEIV YNVSDNRTWA DLTQTLHAFL AGYSPAAQED SINCTSEQYF FQESFRAPNH TKFSCKFTAD MLQNCSGLAD PNFGFEEGKP CFIIKMNRIV KFLPSNGSAP RVDCAFLDQP RELGQPLQVK YYPPNGTFSL HYFPYYGKKA QPHYSNPLVA AKLLNIPRNA EVAIVCKVMA EHVTFNNPHD PYEGKVEFKL KIEK | |
Greater than 90% as determined by SDS-PAGE. |
496 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |