missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPIL1 Protein
A cDNA sequence encoding the PPIL1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10831-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
51645 | |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 300 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin. | |
Human | |
His | |
PPIL1 solution containing 20 mM Tris-HCl buffer (pH 8.0) and 20% glycerol |
PPIL1 Protein | |
Research Use Only | |
PPIL1 | |
Recombinant Protein | |
E. coli | |
MAAIPPDSWQ PPNVYLETSM GIIVLELYWK HAPKTCKNFA ELARRGYYNG TKFHRIIKDF MIQGGDPTGT GRGGASIYGK QFEDELHPDL KFTGAGILAM ANAGPDTNGS QFFVTLAPTQ WLDGKHTIFG RVCQGIGMVN RVGMVETNSQ DRPVDDVKII KAYPSGLEHH HHHH | |
Greater than 95.0% as determined by SDS-PAGE. |