Learn More
enQuireBio™ Recombinant Human PPM1D Protein
A cDNA sequence encoding the PPM1D was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13118-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
PPM1D Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PPM1D protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MVAFFAVCDG HGGREAAQFA REHFWGFIKK QKGFTSSEPA KVCAAIRKGF LACHLAMWKK LAEWPKTMTG LPSTSGTTAS VVIIRGMKMY VAHVGDSGVV LGIQDDPKDD FVRAVEVTQD HKPELPKERE RIEGLGGSVM NKSGVNRVVW KRPRLTHNGP VRRSTVIDQI PFLAVARALG DLWSYDFFSG EFVVSPEPDT SVHTLDPQKH KYIILGSDGL WNMIPPQDAI SMCQDQEEKK YLMGEHGQSC AKMLVNRALG RWRQRMLRAD NTSAIVICI |