missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP1R14A Protein
A cDNA sequence encoding the PPP1R14A was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13123-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
PPP1R14A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAAQRLGKRV LSKLQSPSRA RGPGGSPGGL QKRHARVTVK YDRRELQRRL DVEKWIDGRL EELYRGMEAD MPDEINIDEL LELESEEERS RKIQGLLKSC GKPVEDFIQE LLAKLQGLHR QPGLRQPSPS HDGSLSPLQD RARTAHP | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PPP1R14A protein solution (1 mg/ml) contains 20mM Tris-HCl, pH 8.0, 0.2mM EDTA, 1mM DTT and 10% glycerol. |