Learn More
enQuireBio™ Recombinant Human PRKAR2A Protein
A cDNA sequence encoding the PRKAR2A was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13144-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
PRKAR2A Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PRKAR2A protein solution (0.5 mg/ml) in Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSHIQIP PGLTELLQGY TVEVLRQQPP DLVEFAVEYF TRLREARAPA SVLPAATPRQ SLGHPPPEPG PDRVADAKGD SESEEDEDLE VPVPSRFNRR VSVCAETYNP DEEEEDTDPR VIHPKTDEQR CRLQEACKDI LLFKNLDQEQ LSQVLDAMFE RIVKADEHVI DQGDDGDNFY VIERGTYDIL VTKDNQTRSV GQYDNRGSFG ELALMYNTPR AATIVATSEG SLWGLDRVTF RRIIVKNNAK KRKMFESFIE SVPLLKSLEV SERMKIVDVI GEKIYKDGER IITQGEKADS FYIIESGEVS ILIRSRTKSN KDGGNQEVEI ARCHKGQYFG ELALVTNKPR AASAYAVGDV KCLVMDVQAF ERLLGPCMDI MKRNISHYEE QLVKMFGSSV DLGNLGQ |