Learn More
enQuireBio™ Recombinant Human pro-IGF2 Protein
A cDNA sequence encoding the pro-IGF2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10848-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
pro-IGF2 Protein | |
Research Use Only | |
The specific activity was determined by the dose-dependent stimulation of the proliferation of MCF-7 cells (human breast cancer cell line) and is typically 2-8ng/ml. | |
Human | |
Untagged | |
Greater than 90% as obsereved by SDS-PAGE. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human (HEK293) | |
The Pro-IGF2 was lyophilized in 50mM Sodium Acetate pH 4.5 and 350mM NaCl. |