Learn More
enQuireBio™ Recombinant Human PTP4A2 Protein
A cDNA sequence encoding the PTP4A2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13209-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
PTP4A2 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PTP4A2 (0.5 mg/ml) protein solution contains 20mM Tris-HCl, pH 8.0, 1mM EDTA and 10% Glycerol. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMNRP APVEISYENM RFLITHNPTN ATLNKFTEEL KKYGVTTLVR VCDATYDKAPVEKEGIHVLD WPFDDGAPPP NQIVDDWLNL LKTKFREEPG CCVAVHCVAG LGRAPVLVAL ALIECGMKYE DAVQFIRQKR RGAFNSKQLL YLEKYRPKMR LRFRDTNGHC CVQ |