Learn More
enQuireBio™ Recombinant Human PYCR2 Protein
A cDNA sequence encoding the PYCR2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13221-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
PYCR2 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PYCR2 protein solution (0.25 mg/ml) in phosphate buffered saline (pH 7.4), 50% glycerol, 5mM DTT and 1mM EDTA. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSVGFIG AGQLAYALAR GFTAAGILSA HKIIASSPEM NLPTVSALRK MGVNLTRSNK ETVKHSDVLF LAVKPHIIPF ILDEIGADVQ ARHIVVSCAA GVTISSVEKK LMAFQPAPKV IRCMTNTPVV VQEGATVYAT GTHALVEDGQ LLEQLMSSVG FCTEVEEDLI DAVTGLSGSG PAYAFMALDA LADGGVKMGL PRRLAIQLGA QALLGAAKML LDSEQHPCQL KDNVCSPGGA TIHALHFLES GGFRSLLINA VEASCIRTRE LQSMADQEKI SPAALKKTLL DRVKLESPTV STLTPSSPGK LLTRSLALGG KKD |