missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB13 Protein
A cDNA sequence encoding the RAB13 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13229-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
RAB13 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAKAYDHLFK LLLIGDSGVG KTCLIIRFAE DNFNNTYIST IGIDFKIRTV DIEGKKIKLQ VWDTAGQERF KTITTAYYRG AMGIILVYDI TDEKSFENIQ NWMKSIKENA SAGVERLLLG NKCDMEAKRK VQKEQADKLA REHGIRFFET SAKSSMNVDE AFSSLARDIL LKSGGRRSGN GNKPPSTDLK TCDKKNTNKC | |
Greater than 85.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RAB13 protein (0.25 mg/ml) is supplied in 20mM Tris-HCL, pH 8.0, 1mM DTT, 200mM NaCl, 1mM EDTA and 20% Glycerol. |