Learn More
enQuireBio™ Recombinant Human RALGPS1 Protein
A DNA sequence encoding the Homo sapiens (Human) RALGPS1, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7659-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001177657.1 | |
RALGPS1 Protein | |
1 mg | |
Not determined. | |
Full Length of Isoform 4 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9649 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
RALGPS1 | |
Recombinant Protein | |
E. coli | |
MYKRNGLMAS VLVTSATPQG SSSSDSLEGQ SCDYASKSYD AVVFDVLKVT PEEFASQITL MDIPVFKAIQ PEELASCGWS KKEKHSLAPN VVAFTRRFNQ VSFWVVREIL TAQTLKIRAE ILSHFVKIAK KLLELNNLHS LMSVVSALQS APIFRLTKTW ALLNRKDKTT FEKLDYLMSK EDNYKRTREY IRSLKMVPSI PYLGIYLLDL IYIDSAYPAS GSIMENEQRS NQMNNILRII ADLQVSCSYD HLTTLPHVQK YLKSVRYIEE LQKFVEDDNY KLSLRIEPGS SSPRLVSSKE DLAAM |