Learn More
enQuireBio™ Recombinant Human RBP1 Protein
A cDNA sequence encoding the RBP1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13282-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
RBP1 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RBP1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 20% glycerol. |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDPPAGF VRAGNPAVAA PQSPLSPEGA HFRAAHHPRS TGSRCPGSLQ PSRPLVANWL QSLPEMPVDF TGYWKMLVNE NFEEYLRALD VNVALRKIAN LLKPDKEIVQ DGDHMIIRTL STFRNYIMDF QVGKEFEEDL TGIDDRKCMT TVSWDGDKLQ CVQKGEKEGR GWTQWIEGDE LHLEMRVEGV VCKQVFKKVQ |