missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RRAS Protein
A cDNA sequence encoding the RRAS was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13363-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
RRAS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSSGAASGTG RGRPRGGGPG PGDPPPSETH KLVVVGGGGV GKSALTIQFI QSYFVSDYDP TIEDSYTKIC SVDGIPARLD ILDTAGQEEF GAMREQYMRA GHGFLLVFAI NDRQSFNEVG KLFTQILRVK DRDDFPVVLV GNKADLESQR QVPRSEASAF GASHHVAYFE ASAKLRLNVD EAFEQLVRAV RKYQEQELPP SPPSAPRKKG GGCPC | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RRAS Human solution (0.5 mg/1ml) containing 20mM Tris-HCl pH 8.0, 0.1M NaCl and 10% glycerol. |