missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RWDD1 Protein
A cDNA sequence encoding the RWDD1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13376-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
RWDD1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTDYGEE QRNELEALES IYPDSFTVLS ENPPSFTITV TSEAGENDET VQTTLKFTYS EKYPDEAPLY EIFSQENLED NDVSDILKLL ALQAEENLGM VMIFTLVTAV QEKLNEIVDQ IKTRREEEKK QKEKEAEEAE KQLFHGTPVT IENFLNWKAK FDAELLEIKK KRMKEEEQAG KNKLSGKQLF ETDHNLDTSD IQFLEDAGNN VEVDESLFQE MDDLELEDDE DDPDYNPADP ESDSAD | |
Greater than 85.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RWDD1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 2mM DTT. |