Learn More
enQuireBio™ Recombinant Human SDC2 Protein
A cDNA sequence encoding the SDC2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13438-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
SDC2 Protein | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SDC2 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Insect | |
ADPESRAELT SDKDMYLDNS SIEEASGVYP IDDDDYASAS GSGADEDVES PELTTSRPLP KILLTSAAPK VETTTLNIQN KIPAQTKSPE ETDKEKVHLS DSERKMDPAE EDTNVYTEKH SDSLFKRTEH HHHHH |