missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sentrin-specific protease 8 Protein
A DNA sequence encoding the Homo sapiens (Human) Sentrin-specific protease 8, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7846-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001159812.1 | |
Sentrin-specific protease 8 Protein | |
Research Use Only | |
SENP8 | |
Recombinant Protein | |
E. coli | |
MDPVVLSYMD SLLRQSDVSL LDPPSWLNDH IIGFAFEYFA NSQFHDCSDH VSFISPEVTQ FIKCTSNPAE IAMFLEPLDL PNKRVVFLAI NDNSNQAAGG THWSLLVYLQ DKNSFFHYDS HSRSNSVHAK QVAEKLEAFL GRKGDKLAFV EEKAPAQQNS YDCGMYVICN TEALCQNFFR QQTESLLQLL TPAYITKKRG EWKDLITTLA KK | |
Greater than 90% as determined by SDS-PAGE. |
123228 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |