Learn More
enQuireBio™ Recombinant Human SERTAD2 Protein
A cDNA sequence encoding the SERTAD2 was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP13471-100ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15963679
  • 1740.00€ / 100µg
Expédition estimée: 10-06-2024
Spécification
9792
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMLGKGGK RKFDEHEDGL EGKIVSPCDG PSKVSYTLQR QTIFNISLMK LYNHRPLTEP SLQKTVLINN MLRRIQEELK QEGSLRPMFT PSSQPTTEPS DSYREAPPAF SHLASPSSHP CDLGSTTPLE ACLTPASLLE DDDDTFCTSQ AMQPTAPTKL SPPALLPEKD SFSSALDEIE ELCPTSTSTE AATAATDSVK GTSSEAGTQK LDGPQESRAD DSKLMDSLPG NFEITTSTGF LTDLTLDDIL FADIDTSMYD FDPCTSSSGT ASKMAPVSAD DLLKTLAPYS SQPVTPSQPF KMDLTELDHI MEVLVGS
Greater than 90% as determined by SDS-PAGE.
SERTAD2 Protein
Research Use Only
SERTAD2
Human
His
The SERTAD2 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.