Learn More
enQuireBio™ Recombinant Human SORD Protein
A cDNA sequence encoding the SORD was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10868-25ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
6652 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
SORD | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAAAAKPNNL SLVVHGPGDL RLENYPIPEP GPNEVLLRMH SVGICGSDVH YWEYGRIGNF IVKKPMVLGH EASGTVEKVG SSVKHLKPGD RVAIEPGAPR ENDEFCKMGR YNLSPSIFFC ATPPDDGNLC RFYKHNAAFC YKLPDNVTFE EGALIEPLSV GIHACRRGGV TLGHKVLVCG AGPIGMVTLL VAKAMGAAQV VVTDLSATRL SKAKEIGADL VLQISKESPQ EIARKVEGQL GCKPEVTIEC TGAEASIQAG IYATRSGGTL VLVGLGSEMT TVPLLHAAIR EVDIKGVFRY CNTWPVAISM LASKSVNVKP LVTHRFPLEK ALEAFETFKK GLGLKIMLKC DPSDQNP |
SORD Protein | |
25 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 0.2 units/mg, in which one unit will convert 1.0 umole of D-fructose to D-sorbitol per minute at pH 7.5 at 25C. | |
Human | |
His | |
SORD protein solution (0.5 mg/ml) is formulated in 20mM Tris-HCl pH 8.0, 0.2M NaCl, 5mM DTT and 20% glycerol. |