Learn More
enQuireBio™ Recombinant Human Squalene synthase Protein
A DNA sequence encoding the Homo sapiens (Human) Squalene synthase, was expressed in the hosts and tags indicated.
Marque:  enQuireBio™ QP6031-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15999489
  • 2285.00€ / 1mg
Expédition estimée: 10-05-2024
Spécification
NP_001274671.1
Squalene synthase Protein
Research Use Only
FDFT1
Recombinant Protein
E. coli
EFVKCLGHPE EFYNLVRFRI GGKRKVMPKM DQDSLSSSLK TCYKYLNQTS RSFAAVIQAL DGEMRNAVCI FYLVLRALDT LEDDMTISVE KKVPLLHNFH SFLYQPDWRF MESKEKDRQV LEDFPTISLE FRNLAEKYQT VIADICRRMG IGMAEFLDKH VTSEQEWDKY CHYVAGLVGI GLSRLFSASE FEDPLVGEDT ERANSMGLFL QKTNIIRDYL EDQQGGREFW PQEVWSRYVK KLGDFAKPEN IDLAVQCLNE LITNALHHIP DVITYLSRLR NQSVFNFCAI PQVMAIATLA ACYNNQQVFK GAVKIRKGQA VTLMMDATNM PAVKAIIYQY MEEIYHRIPD SDPSSSKTRQ IISTIRTQNL PNCQLISRSH YSPIYLSFVM LLAALSWQYL TTLSQVTEDY VQTGEH
Greater than 90% as determined by SDS-PAGE.
2222
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol