Learn More
enQuireBio™ Recombinant Human STAMBPL1 Protein
A cDNA sequence encoding the STAMBPL1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13607-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
STAMBPL1 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The STAMBPL1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl and 20% glycerol. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSQ SHMDQPFTVN SLKKLAAMPD HTDVSLSPEE RVRALSKLGC NITISEDITP RRYFRSGVEM ERMASVYLEE GNLENAFVLY NKFITLFVEK LPNHRDYQQC AVPEKQDIMK KLKEIAFPRT DELKNDLLKK YNVEYQEYLQ SKNKYKAEIL KKLEHQRLIE AERKRIAQMR QQQLESEQFL FFEDQLKKQE LARGQMRSQQ TSGLSEQIDG SALSCFSTHQ NNSLLNVFAD QPNKSDATNY ASHSPPVNRA LTPAATLSAV QNLVVEGLRC VVLPEDLCHK FLQLAESNTV RGIETCGILC GKLTHNEFTI THVIVPKQSA GPDYCDMENV EELFNVQDQH DLLTLGWIHT HPTQTAFLSS VDLHTHCSYQ LMLPEAIAIV CSPKHKDTGI FRLTNAGMLE VSACKKKGFH PHTKEPRLFS ICKHVLVKDI KIIVLDLR |