missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Steroid 17-alpha-hydroxylase / 17,20 lyase Protein
A DNA sequence encoding the Homo sapiens (Human) Steroid 17-alpha-hydroxylase/17,20 lyase, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5904-ec-200ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_000093.1 | |
Steroid 17-alpha-hydroxylase / 17,20 lyase Protein | |
Research Use Only | |
CYP17A1 | |
Recombinant Protein | |
E. coli | |
MWELVALLLL TLAYLFWPKR RCPGAKYPKS LLSLPLVGSL PFLPRHGHMH NNFFKLQKKY GPIYSVRMGT KTTVIVGHHQ LAKEVLIKKG KDFSGRPQMA TLDIASNNRK GIAFADSGAH WQLHRRLAMA TFALFKDGDQ KLEKIICQEI STLCDMLATH NGQSIDISFP VFVAVTNVIS LICFNTSYKN GDPELNVIQN YNEGIIDNLS KDSLVDLVPW LKIFPNKTLE KLKSHVKIRN DLLNKILENY KEKFRSDSIT NMLDTLMQAK MNSDNGNAGP DQDSELLSDN HILTTIGDIF GAGVETTTSV VKWTLAFLLH NPQVKKKLYE EIDQNVGFSR TPTISDRNRL LLLEATIREV LRLRPVAPML IPHKANVDSS IGEFAVDKGT EVIINLWALH HNEKEWHQPD QFMPERFLNP AGTQLISPSV SYLPFGAGPR SCIGEILARQ ELFLIMAWLL QRFDLEVPDD GQLPSLEGIP KVVFLIDSFK VKIKVRQAWR EAQAEGST | |
Greater than 90% as determined by SDS-PAGE. |
1586 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |