missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STX2 Protein

A cDNA sequence encoding the STX2 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13634-2ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15924219

  • 137.00€ / 2µg
Expédition estimée: 04-07-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

STX2 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The STX2 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 50% glycerol.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMMRDRL PDLTACRKND DGDTVVVVEK DHFMDDFFHQ VEEIRNSIDK ITQYVEEVKK NHSIILSAPN PEGKIKEELE DLNKEIKKTA NKIRAKLKAI EQSFDQDESG NRTSVDLRIR RTQHSVLSRK FVEAMAEYNE AQTLFRERSK GRIQRQLEIT GRTTTDDELE EMLESGKPSI FTSDIISDSQ ITRQALNEIE SRHKDIMKLE TSIRELHEMF MDMAMFVETQ GEMINNIERN VMNATDYVEH AKEETKKAIK YQSKARRKK
Vidéos
FDS
Documents
Certificats
Promotions

Promotions