Learn More
enQuireBio™ Recombinant Human THG1L Protein
A cDNA sequence encoding the THG1L was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP13742-2ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15924559
  • 131.00€ / 2µg
Expédition estimée: 21-06-2024
Spécification
54974
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAKSKFE YVRDFEADDT CLAHCWVVVR LDGRNFHRFA EKHNFAKPND SRALQLMTKC AQTVMEELED IVIAYGQSDE YSFVFKRKTN WFKRRASKFM THVASQFASS YVFYWRDYFE DQPLLYPPGF DGRVVVYPSN QTLKDYLSWR QADCHINNLY NTVFWALIQQ SGLTPVQAQG RLQGTLAADK NEILFSEFNI NYNNELPMYR KGTVLIWQKV DEVMTKEIKL PTEMEGKKMA VTRTRTKPVP LHCDIIGDAF WKEHPEILDE DS
Greater than 90% as determined by SDS-PAGE.
THG1L Protein
Research Use Only
THG1L
Human
His
The THG1L solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 20% glycerol.