missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNF receptor-associated factor 5 Protein
A DNA sequence encoding the Homo sapiens (Human) TNF receptor-associated factor 5, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6837-ec-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001029082.1 | |
TNF receptor-associated factor 5 Protein | |
Research Use Only | |
TRAF5 | |
Recombinant Protein | |
E. coli | |
MAYSEEHKGM PCGFIRQNSG NSISLDFEPS IEYQFVERLE ERYKCAFCHS VLHNPHQTGC GHRFCQHCIL SLRELNTVPI CPVDKEVIKS QEVFKDNCCK REVLNLYVYC SNAPGCNAKV ILGRYQDHLQ QCLFQPVQCS NEKCREPVLR KDLKEHLSAS CQFRKEKCLY CKKDVVVINL QNHEENLCPE YPVFCPNNCA KIILKTEVDE HLAVCPEAEQ DCPFKHYGCA VTDKRRNLQQ HEHSALREHM RLVLEKNVQL EEQISDLHKS LEQKESKIQQ LAETIKKLEK EFKQFAQLFG KNGSFLPNIQ VFASHIDKSA WLEAQVHQLL QMVNQQQNKF DLRPLMEAVD TVKQKITLLE NNDQRLAVLE EETNKHDTHI NIHKAQLSKN EERFKLLEGT CYNGKLIWKV TDYKMKKREA VDGHTVSIFS QSFYTSRCGY RLCARAYLNG DGSGRGSHLS LYFVVMRGEF DSLLQWPFRQ RVTLMLLDQS GKKNIMETFK PDPNSSSFKR PDGEMNIASG CPRFVAHSVL ENAKNAYIKD DTLFLKVAVD LTDLEDL | |
Greater than 90% as determined by SDS-PAGE. |
7188 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |