missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFRSF14 Protein
A cDNA sequence encoding the TNFRSF14 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10889-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
8740 | |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Fully biologically active when compared to standard. The biologically active as determined by its ability to inhibit TNF-beta -mediated cytotoxicity using murine L929 cells. | |
Human | |
Untagged | |
TNFRSF14 protein was lyophilized from a 0.2 microm filtered concentrated solution in PBS, pH 7.4, with 3 % Trehalose. |
TNFRSF14 Protein | |
Research Use Only | |
TNFSF14 | |
Recombinant Protein | |
P. pastoris | |
LPSCKEDEYP VGSECCPKCS PGYRVKEACG ELTGTVCEPC PPGTYIAHLN GLSKCLQCQM CDPAMGLRAS RNCSRTENAV CGCSPGHFCI VQDGDHCAAC RAYATSSPGQ RVQKGGTESQ DTLCQNCPPG TFSPNGTLEE CQHQTKCSWL VTKAGAGTSS SHWVEPKSSD KTHTCPPCPA PEFEGAPSVF LFPPKPKDTL MISRTPEVTC VVVDVSHEDP EVKFNWYVDG VEVHNAKTKP REEQYNSTYR VVSVLTVLHQ DWLNGKEYKC KVSNKALPTP IEKTISKAKG QPREPQVYTL PPSRDELTKN QVSLTCLVKG FYPSDIAVEW ESNGQPENNY KTTPPVLDSD GSFFLYSKLT VDKSRWQQGN VFSCSVMHEA LHNHYTQKSL SLSPGK | |
Greater than 95.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |