Learn More
enQuireBio™ Recombinant Human Twinfilin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Twinfilin-2, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7777-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_009215.1 | |
Twinfilin-2 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
11344 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
TWF2 | |
Recombinant Protein | |
E. coli | |
AHQTGIHATE ELKEFFAKAR AGSVRLIKVV IEDEQLVLGA SQEPVGRWDQ DYDRAVLPLL DAQQPCYLLY RLDSQNAQGF EWLFLAWSPD NSPVRLKMLY AATRATVKKE FGGGHIKDEL FGTVKDDLSF AGYQKHLSSC AAPAPLTSAE RELQQIRINE VKTEISVESK HQTLQGLAFP LQPEAQRALQ QLKQKMVNYI QMKLDLERET IELVHTEPTD VAQLPSRVPR DAARYHFFLY KHTHEGDPLE SVVFIYSMPG YKCSIKERML YSSCKSRLLD SVEQDFHLEI AKKIEIGDGA ELTAEFLYDE VHPKQHAFKQ AFAKPKGPGG KRGHKRLIRG PGENGDDS |