Learn More
enQuireBio™ Recombinant Human TYRP1 Protein
A cDNA sequence encoding the TYRP1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13846-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
TYRP1 Protein | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TYRP1 protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Insect | |
ADLQFPRQCA TVEALRSGMC CPDLSPVSGP GTDRCGSSSG RGRCEAVTAD SRPHSPQYPH DGRDDREVWP LRFFNRTCHC NGNFSGHNCG TCRPGWRGAA CDQRVLIVRR NLLDLSKEEK NHFVRALDMA KRTTHPLFVI ATRRSEEILG PDGNTPQFEN ISIYNYFVWT HYYSVKKTFL GVGQESFGEV DFSHEGPAFL TWHRYHLLRL EKDMQEMLQE PSFSLPYWNF ATGKNVCDIC TDDLMGSRSN FDSTLISPNS VFSQWRVVCD SLEDYDTLGT LCNSTEDGPI RRNPAGNVAR PMVQRLPEPQ DVAQCLEVGL FDTPPFYSNS TNSFRNTVEG YSDPTGKYDP AVRSLHNLAH LFLNGTGGQT HLSPNDPIFV LLHTFTDAVF DEWLRRYNAD ISTFPLENAP IGHNRQYNMV PFWPPVTNTE MFVTAPDNLG YTYEIQWPSR EFSVPEHHHH HH |