Learn More
enQuireBio™ Recombinant Human UTI Protein
A cDNA sequence encoding the UTI was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10909-10mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
UTI Protein | |
Research Use Only | |
Human UTI has an activity of 2390IU/mg. | |
Human | |
Untagged |
10 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Native Protein | |
Lyophilized from a (1 mg/ml) solution containing no additives. |