Learn More
enQuireBio™ Recombinant Human XRCC2 Protein
A cDNA sequence encoding the XRCC2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13973-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
XRCC2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMCSAFHR AESGTELLAR LEGRSSLKEI EPNLFADEDS PVHGDILEFH GPEGTGKTEM LYHLTARCIL PKSEGGLEVE VLFIDTDYHF DMLRLVTILE HRLSQSSEEI IKYCLGRFFL VYCSSSTHLL LTLYSLESMF CSHPSLCLLI LDSLSAFYWI DRVNGGESVN LQESTLRKCS QCLEKLVNDY RLVLFATTQT IMQKASSSSE EPSHASRRLC DVDIDYRPYL CKAWQQLVKH RMFFSKQDDS QSSNQFSLVS RCLKSNSLKK HFFIIGESGV EFC | |
Greater than 80% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The XRCC2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |