missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ZMAT3 Protein

A cDNA sequence encoding the ZMAT3 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP14002-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15915389

  • 205.00€ / 10µg
Expédition estimée: 30-06-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

64393
Greater than 85% as determined by SDS-PAGE.
Research Use Only
ZMAT3
Human
His
The ZMAT3 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM DTT and 50% glycerol.
ZMAT3 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMILLQHA VLPPPKQPSP SPPMSVATRS TGTLQLPPQK PFGQEASLPL AGEEELSKGG EQDCALEELC KPLYCKLCNV TLNSAQQAQA HYQGKNHGKK LRNYYAANSC PPPARMSNVV EPAATPVVPV PPQMGSFKPG GRVILATEND YCKLCDASFS SPAVAQAHYQ GKNHAKRLRL AEAQSNSFSE SSELGQRRAR KEGNEFKMMP NRRNMYTVQN NSAGPYFNPR SRQRIPRDLA MCVTPSGQFY CSMCNVGAGE EMEFRQHLES KQHKSKVSEQ RYRNEMENLG YV
Vidéos
FDS
Documents
Certificats
Promotions

Promotions