missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H1N1 Taiwan Protein
Recombinant Protein
Marque: enQuireBio™ QP12079-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
Influenza H1N1 Taiwan Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Insect | |
The H1N1A/Taiwan/1/86 solution contains STE, 0.1% sodium azide (NaN3) and 0.005% thimerosal. |